Learn More
Sperian™ Honeywell™ Clearways™ CB20 Browguard with Ratchet Headband
Product Code. 12622733
Change view
Click to view available options
Weight (Metric):
94g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
12622733 94g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12622733 Supplier Sperian™ Supplier No. 1002341
  • ONLINE EXCLUSIVE PRICE
    42.42€
    18.78€ / Pack of 1
    Save 23.64€
    55% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
Offering both strength and durability with economical replacement visors available in polycarbonate. Sperian™ Honeywell™ Clearways™ CB20 Browguards with Ratchet Headbands are lightweight and comfortable enough to wear all day.
  • Recommended for the agriculture, chemical and forestry industries
  • Protects against hazards including abrasion, airborne particles, biohazards, chemicals, impacts, splashes and more
  • Visors available in polycarbonate
  • **Face shield is not included**
TRUSTED_SUSTAINABILITY
Specifications
Product Type Browguard
Weight (Metric) 94g