Learn More
Tenak™ Classic Rack for Chest Freezers, with Locking Rod, 32 mm Box height
Classic Rack is suitable for use with all chest freezer models.
80.10€ - 95.40€
Specifications
Material | Stainless Steel |
---|---|
For Use With (Equipment) | Chest Frseezers |
Includes | Locking rod |
Height (Metric) | 32 mm |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15200474
|
Tenak™
TE21354 |
Boxes (12) |
87.45€
Each |
Estimated Shipment:
Log in to see stock. |
|||||
15230474
|
Tenak™
TE21358 |
Boxes (16) |
95.40€
Each |
Estimated Shipment:
Log in to see stock. |
|||||
15290464
|
Tenak™
TE21350 |
Boxes (8) |
80.10€
Each |
Estimated Shipment:
Log in to see stock. |
|||||
Description
- Comes with a locking rod to secure the cryoboxes
- Measure the inside height of you freezer when choosing the right rack
- Constructed of stainless steel
Specifications
Stainless Steel | |
Chest Frseezers | |
Locking rod | |
32 mm |