Learn More
Abeba™ Classic 1011 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
Brand:  Abeba™ 1011/43
Additional Details : Weight : 0.88600kg
 View more versions of this product
Product Code. 15616597
  • 88.22€ / 1 pair
Estimated Shipment: 04-06-2024
Description
  • CE EN ISO 20345:2011 SB A, E, FO SRC
  • Microfiber black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Microfiber lining and heel patch
  • Adjustable instep area
  • Buckle fastening
Specifications
Specifications
CE EN ISO 20345:2011 SB A, E, FO SRC
Microfiber
43
Polyurethane
Steel
Black
Microfiber
Slip-resistant
1011
Animal fats, oil and fuel