Learn More
Abeba™ Classic 1010 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
Brand:  Abeba™ 1010/43
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15616587
  • 80.64€ / 1 pair
Estimated Shipment: 31-05-2024
Description
  • CE EN ISO 20345:2011 SB A, E, FO SRC
  • Grained leather black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Adjustable instep area
  • Buckle fastening
  • Heel patch microfiber
Specifications
Specifications
CE EN ISO 20345:2011 SB A, E, FO SRC
Microfiber
43
Polyurethane
Steel
Black
Leather
Slip-resistant
1010
Animal fats, oil and fuel