Learn More
Hirschmann™ Class B Measuring Cylinders, Blue Graduation, with Plastic Socket and Protection Collar
Product Code. 10022641
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10022641 50 mL Pack of 2
10607315 10 mL Pack of 2
12521467 25 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10022641 Supplier Hirschmann™ Supplier No. 2260175
Add to basket
Add to basket
Excellent for precise liquid measurement and fluid transfer in the laboratory. Manufactured from durable DURAN™ glass tubing.
  • With spout, plastic socket and protection collar, short line graduation
  • Blue graduation
  • Class B
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications
Material Glass
Capacity (Metric) 50 mL
Graduations 1 mL
Autoclavable Autoclavable
Subdivision 1 mL
Description Calibration To Contain, with plastic socket and protection collar
Sterility Non-sterile
Calibration To Contain
Base Type Hexagonal
Scale Color Blue
Shape Round
Class Class B
Height (Metric) 200 mm
Certifications/Compliance DIN EN ISO 4788
Tolerance ±0.75 mL
Product Type Graduated Measuring Cylinder