Learn More
Hirschmann™ Class AS AR™ Glass Volumetric Pipettes
Product Code. 11762162
Change view
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2 mL
3 mL
5 mL
10 mL
15 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Pack of 12
Pack of 6
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11762162 20 mL Pack of 6
11792162 100 mL Pack of 6
11772252 0.5 mL Pack of 12
11742162 10 mL Pack of 6
11752162 15 mL Pack of 6
11722152 1 mL Pack of 12
11772162 25 mL Pack of 6
11732152 2 mL Pack of 12
11722162 3 mL Pack of 6
11782162 50 mL Pack of 6
11732162 5 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 11762162 Supplier Hirschmann™ Supplier No. 1340265
  • ONLINE EXCLUSIVE PRICE
    97.00€
    86.10€ / Pack of 6
    Save 10.90€
    11% Off
In Stock
Add to basket
Edge
Add to basket
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS AR™ Glass Volumetric Pipettes come with amber stain graduation marks.
  • Acc. to DIN EN ISO 648
  • Volume between two marks
  • Conformity certified, with dated batch identification
Specifications
Class AS
Material AR-GLAS
Graduated Yes
Graduation Color Brown
Quantity 6/Pk.
Tolerance 0.03 mL
Volume (Metric) 20 mL
Length (Metric) 520 mm
Type Volumetric Pipet