Learn More
- Home
- Products
- Stoppers and Closures
- Stoppers
- Fisherbrand™ Cellulose Stoppers
Fisherbrand™ Cellulose Stoppers
Product Code. 11798523 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Bottom:
6 mm
6.5 mm
7 mm
8 mm
10 mm
11 mm
12 mm
13 mm
15 mm
16 mm
18 mm
19 mm
23 mm
24 mm
27 mm
29 mm
30 mm
35 mm
37 mm
40 mm
Length (Metric):
11 mm
30 mm
32 mm
35 mm
37 mm
40 mm
42 mm
43 mm
50 mm
60 mm
65 mm
Unit Size:
Pack of 100
Pack of 1000
Pack of 200
Pack of 50
Pack of 500
25 product options available for selection
| Product Code. | Diameter (Metric) Bottom | Length (Metric) | Unit Size |
|---|---|---|---|
| 11798523 | 27 mm | 60 mm | Pack of 200 |
| 11768513 | 7 mm | 30 mm | Pack of 1000 |
| 11740096 | 11 mm | 32 mm | Pack of 1000 |
| 11750096 | 11 mm | 40 mm | Pack of 1000 |
| 11778513 | 8 mm | 35 mm | Pack of 1000 |
| 11760096 | 12 mm | 37 mm | Pack of 1000 |
| 11788513 | 10 mm | 37 mm | Pack of 1000 |
| 11758533 | 12 mm | 42 mm | Pack of 1000 |
| 11770096 | 12 mm | 32 mm | Pack of 1000 |
| 11798513 | 15 mm | 30 mm | Pack of 1000 |
| 11708523 | 16 mm | 30 mm | Pack of 1000 |
| 11748513 | 13 mm | 32 mm | Pack of 1000 |
| 11768523 | 19 mm | 30 mm | Pack of 1000 |
| 11758523 | 18 mm | 30 mm | Pack of 1000 |
| 11778523 | 23 mm | 30 mm | Pack of 500 |
| 11784506 | 24 mm | 43 mm | Pack of 200 |
| 11788523 | 27 mm | 60 mm | Pack of 200 |
| 11708533 | 29 mm | 60 mm | Pack of 200 |
| 11728533 | 30 mm | 40 mm | Pack of 200 |
| 11718533 | 35 mm | 40 mm | Pack of 200 |
| 11738533 | 35 mm | 60 mm | Pack of 100 |
| 11720015 | 37 mm | 50 mm | Pack of 100 |
| 11721666 | 40 mm | 65 mm | Pack of 50 |
| 11730096 | 6 mm | 11 mm | Pack of 1000 |
| 11758513 | 6.5 mm | 30 mm | Pack of 1000 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
Specifications
Specifications
| Disposable | Single-use |
| For Use With (Application) | Beakers, Erlenmeyer flasks, tubes, and different types of bottles |
| Length (Metric) | 60 mm |
| Quantity | 200PK |
| Temperature Range (Metric) | 200°C |
| Diameter (Metric) Bottom | 27 mm |
| Diameter (Metric) Top | 36 mm |
| Material | Cellulose Fiber |
| Product Type | Stopper |
| Sterility | Sterile |