Learn More
Fisherbrand™ Cellulose Stoppers
For flasks, erlenmeyers and test tubes.
Marke: Fisherbrand™
Weitere Details : Weight : 0.01000kg
Beschreibung
- Permeable in air
- Sterilizable to 200°C maximum
- Single use
Spezifikation
Yes | |
Stopper | |
11 mm | |
1000PK | |
200°C |
Sterile | |
Beakers, Erlenmeyer flasks, tubes, and different types of bottles | |
17 mm | |
Cellulose Fiber | |
40 mm |