Learn More
Proteintech ChromoTek RFP-Trap™ Magnetic Agarose
The ChromoTek RFP-Trap™ Magnetic Agarose are affinity beads for IP of RFP-fusion proteins.
Brand:  Proteintech RTMA-100
Additional Details : Weight : 0.00120kg
 View more versions of this product
Product Code. 17353453
  • 1798.00€ / 100 reactions
Estimated Shipment: 05-08-2024
Description
  • Comprises a RFP Nanobody/ VHH coupled to magnetic agarose
Specifications
Specifications
Upon receipt store at 4°C. Do not freeze.
mRFP, mCherry, mRFPruby, mPlum, tagRFP, mKate2, mOrange, PA-mCherry, mScarlet
Alpaca
IP, CoIP, ChIP, RIP
Magnetic Agarose
100 Reactions