Learn More
Proteintech ChromoTek pSpot8 GFP-Spot-Tag
pSpot8 GFP-Spot-Tag™ vector for expression of GFP-Spot-Tag™ fusion protein in S. cerevisiae - positive control.
Brand: Proteintech EV-33
Additional Details : Weight : 0.00240kg
Description
- The plasmid pSpot8_GFP-Spot-Tag is a S. cerevisiae expression vector encoding the Aequorea victoria Green fluorescent protein (GFP) variant GFPuv fused to the Spot-Tag™. The Spot-Tag™ comprises the sequence PDRVRAVSHWSS and is fused to the C-terminus of GFPuv. This plasmid may be used as a positive control for immunoprecipitation or Western blot experiments using the Spot-Tag™ system.
Specifications
10 μg |