Learn More
Proteintech ChromoTek pSpot2 GFPSpot-Tag Vector
pSpot2 GFP-Spot-Tag™ vector for expression of GFP-Spot-Tag™ fusion protein in E. coli - positive control.
Brand: Proteintech EV-32
Additional Details : Weight : 0.00240kg
Description
- The plasmid pSpot2_GFP is a bacterial expression vector encoding Aequorea victoria Enhanced Green fluorescent protein (EGFP) fused to the Spot-Tag™. The Spot-Tag™ comprises the sequence PDRVRAVSHWSS and is fused to the C-terminus of EGFP. This plasmid may be used as a positive control for immunoprecipitation or Western blot experiments using the Spot-Tag™ system.
Specifications
10 μg |