Learn More
Proteintech ChromoTek pSpot2 GFPSpot-Tag Vector
pSpot2 GFP-Spot-Tag™ vector for expression of GFP-Spot-Tag™ fusion protein in E. coli - positive control.
Brand:  Proteintech EV-32
Additional Details : Weight : 0.00240kg
Product Code. 17373323
  • 94.00€ / 10µg
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • The plasmid pSpot2_GFP is a bacterial expression vector encoding Aequorea victoria Enhanced Green fluorescent protein (EGFP) fused to the Spot-Tag™. The Spot-Tag™ comprises the sequence PDRVRAVSHWSS and is fused to the C-terminus of EGFP. This plasmid may be used as a positive control for immunoprecipitation or Western blot experiments using the Spot-Tag™ system.