Learn More
Proteintech ChromoTek GFP-Trap™ Magnetic Particles M-270
GFP-Trap™ Magnetic Particles M-270 for immunoprecipitation (IP) of GFP-tagged proteins.
Brand: Proteintech GTD-200
Additional Details : Weight : 0.00240kg
Description
- GFP-Trap™ Magnetic Particles M-270 is highly recommended, when very large proteins/complexes are investigated, and magnetic separation is needed for IP
- Consists of a GFP VHH/ Nanobody coupled to Magnetic Particles M-270
Specifications
10 μL slurry bind about 1 μg of recombinant GFP | |
IP, CoIP, ChIP, RIP | |
Magnetic Particles M-270 | |
200 Reactions |
Upon receipt store at 4°C. Do not freeze. | |
AcGFP, Clover, eGFP, Emerald, GFP, GFP5, GFP Envy, GFP S65T, mGFP, mPhluorin, PA-GFP, Superfolder GFP, TagGFP, TagGFP2, monomeric eGFP A206K, CFP, YFP, Citrine, eCitrine, eYFP, Venus, Ypet, BFP | |
Alpaca |