missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek GFP-Booster Alexa Fluor™ 647
GFP VHH conjugated to Alexa Fluor™ 647.
171.00€ - 364.00€
Specifications
Clonality | Monoclonal |
---|---|
For Use With (Application) | Immunofluorescence (IF), Immunohistochemistry (IHC), Immunocytochemistry (ICC) |
Conjugate | Alexa Fluor™ 647 |
Target Specificity | AcGFP, Citrine, CFP, eGFP, eYFP, GFP S65T, pHluorin, sfGFP, mClover (Clover A206K), TagGFP, tagGFP2, Venus, wtGFP, YFP |
Content And Storage | Shipped at ambient temperature. Store at -20°C/-4°F. Avoid freeze-thaw cycles. Aliquot upon arrival. Protect from light. Stable for 6 months. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17383333
|
Proteintech
GB2AF647-10 |
10 μL |
171.00€
10µL |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
17393333
|
Proteintech
GB2AF647-50 |
50 μL |
364.00€
50µL |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Description
- anti-GFP VHH/ Nanobody conjugated to a fluorescent dye for IF/ microscopy
Specifications
Monoclonal | |
Alexa Fluor™ 647 | |
Shipped at ambient temperature. Store at -20°C/-4°F. Avoid freeze-thaw cycles. Aliquot upon arrival. Protect from light. Stable for 6 months. |
Immunofluorescence (IF), Immunohistochemistry (IHC), Immunocytochemistry (ICC) | |
AcGFP, Citrine, CFP, eGFP, eYFP, GFP S65T, pHluorin, sfGFP, mClover (Clover A206K), TagGFP, tagGFP2, Venus, wtGFP, YFP |