Learn More
Macherey-Nagel™ Chromafix™ SA Cartridge
Brand: Macherey-Nagel™ 731833
Additional Details : Weight : 0.01000kg
Specifications
Silica | |
Amino Acids, Amines, Chlorophyll, PCBs | |
SA | |
60 Å | |
500 m2/g |
Adsorbent Weight: 920 mg | |
45 μm | |
Chromafix SA Cartridge | |
50 Pack |