Learn More
Bryson Packaging™ Resealable Bags
Description
- Clear in color
- Tight seal ensured
Specifications
Specifications
| Description | Resealable Bag |
| Color | Clear |
| For Use With (Application) | Ideal for storing stationery and sending samples |
| Material | Polyethylene |
| Quantity | 100 Pack |
| Height (Metric) | 280 mm |
| Product Type | Sample Bag |
| Width (Metric) | 205 mm |