Learn More
WTW™ Chloride Test Kits
Get accurate water test results for waste water, drinking water, ground water, and mineral water/after with chromate reagent test kits.
Brand: WTW™ 250491
Additional Details : Weight : 0.30000kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Description
- Variety of instrumentation for measuring chloride
- Chromate reagent test kit
- For waste water, drinking water, ground water, mineral water/after dilution
- PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, Photoflex photometers
Specifications
DIN 38405 D31 | |
5 mL | |
PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer, Photoflex™ meter | |
2.5 to 250 mg/L | |
Indophenol blue (Berthelot reaction) |
13 at 20°C | |
Wastewater, drinking water, ground water, mineral water/after dilution | |
Ammonium | |
2°C to 8°C | |
CombiCheck Ammonium (NH4) Reagent Test Kit |
Safety and Handling
- chloride test method
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : iron (III) nitrate,nitric acid