Learn More
Fisherbrand™ Chest ULT Freezer Racks
Product Code. 15709539 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
12 Boxes (2 in.)
2 in. Boxes (10)
2 in. Boxes (11)
2 in. Boxes (13)
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
15709539 2 in. Boxes (13) Each
15779529 2 in. Boxes (10) Each
15789529 2 in. Boxes (11) Each
15799529 12 Boxes (2 in.) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15709539
  • ONLINE EXCLUSIVE PRICE
    56.39€
    49.09€ / Pack of 1
    Save 7.30€
    12% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
Fisherbrand inventory control racks are ideal for all cold storage application. They fit all standard types of chest ULT freezers.
  • Holds standard cryoboxes
  • Made of high quality stainless steel
  • Locking rod included
Specifications
Holds 2 in. Boxes (13)
Material Stainless Steel
For Use With (Application) Standard 2 in. Cryoboxes
For Use With (Equipment) Chest ULT Freezer
Dimensions (L x W x H) 717 x 143 x 140 mm
Product Type Vertical Rack