Learn More
Fisherbrand™ Chemical Absorbent Pads
Product Code. 15853720 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
89 L
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15853720 89 L Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15853720
  • ONLINE EXCLUSIVE PRICE
    222.00€
    119.95€ / Pack of 100
    Save 102.05€
    45% Off
In Stock
Add to basket
Edge
Add to basket
Fisherbrand™ Chemical Absorbent Pads absorb chemicals and hazardous liquids
  • One sided spunbound gives extra strength as well as less lint
  • Does not degrade or cause a dangerous chemical reaction with the absorbed liquid
  • Green color alerts workers that the product is being used for hazardous liquids and allows the user to identify and separate hazardous waste for disposal, resulting in lower disposal costs
  • Perforated for more flexibility - use only what you need and minimize waste
  • Perfect to clean up chemical spills or for use in laboratories
Specifications
Color Green
Description Pads, 50 x 40 cm, Medium-weight, Perforated and bonded
Capacity (Metric) 89 L
Material Spunbond, Meltblown
Type Chemical Sorbent
For Use With (Application) Overspray, leaks and drips in drum and liquid storage areas, workstations and laboratories
Packaging Quantity 100 Pads/Cs.
Thickness 2-ply
Dimensions (L x W) 50 x 40 cm
Length (Metric) 50 cm
Width (Metric) 40 cm
Height (English) 16.14 in.
Weight (English) 15.65 lb.
Weight (Metric) 7.1 kg