Learn More
Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Flasks
Ideal for low-serum, serum-free cultivation and cell differentiation. Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Flasks helps in promotion of cell adhesion and proliferation
Brand: Greiner Bio-One 690940
Additional Details : Weight : 0.80000kg
Description
Poly-D-lysine (PDL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.- Coated with Poly-D-Lysine synthetic protein (75-150 kDa)
- For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
- Ready-to-use products
- Includes lid with condensation rings
Specifications
Poly-D-Lysine | |
Plug Seal Cap | |
25 cm2 | |
50 mL |