Learn More
Cefmetazole, Thermo Scientific™
A broad spectrum second generation cephalosporin antibiotic
367.00€
Chemical Identifiers
| CAS | 56796-20-4 |
|---|---|
| Molecular Formula | C15H17N7O5S3 |
| Molecular Weight (g/mol) | 471.53 |
| MDL Number | MFCD00865068 |
| InChI Key | SNBUBQHDYVFSQF-HIFRSBDPSA-N |
| PubChem CID | 42008 |
| ChEBI | CHEBI:3489 |
| IUPAC Name | (6R,7S)-7-{2-[(cyanomethyl)sulfanyl]acetamido}-7-methoxy-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
| SMILES | CO[C@]1(NC(=O)CSCC#N)[C@H]2SCC(CSC3=NN=NN3C)=C(N2C1=O)C(O)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 56796-20-4 | |
| 471.53 | |
| SNBUBQHDYVFSQF-HIFRSBDPSA-N | |
| CHEBI:3489 | |
| CO[C@]1(NC(=O)CSCC#N)[C@H]2SCC(CSC3=NN=NN3C)=C(N2C1=O)C(O)=O |
| C15H17N7O5S3 | |
| MFCD00865068 | |
| 42008 | |
| (6R,7S)-7-{2-[(cyanomethyl)sulfanyl]acetamido}-7-methoxy-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
Specifications
| 56796-20-4 | |
| C15H17N7O5S3 | |
| Soluble in water | |
| CO[C@]1(NC(=O)CSCC#N)[C@H]2SCC(CSC3=NN=NN3C)=C(N2C1=O)C(O)=O | |
| 471.53 | |
| CHEBI:3489 | |
| Cefmetazole |
| White | |
| MFCD00865068 | |
| SNBUBQHDYVFSQF-HIFRSBDPSA-N | |
| (6R,7S)-7-{2-[(cyanomethyl)sulfanyl]acetamido}-7-methoxy-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid | |
| 42008 | |
| 471.53 |
Safety and Handling
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
missing translation for 'einecsNumber' : 260-384-2
missing translation for 'rtecsNumber' : XI0372000
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.