Learn More
Abnova™ CDK7/CEN5q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0181.100uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16033285
  • 1475.00€ / 100µL
Estimated Shipment:
to see stock.
Add to basket
Specifications
Store at 4°C in the dark.
Tube
1022
100 μL
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
FISH-Ce
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe