Learn More
IBA Lifesciences CD8 Fab-Strep
Low affinity Fab fragment fused to a Twin-Strep-tag™
Brand:  IBA Lifesciences 6-8505-150
Additional Details : Weight : 0.00600kg
 View more versions of this product
Product Code. 17227610
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CD8 Fab-Strep is a recombinantly expressed low affinity Fab fragment fused to a Twin-Strep-tag™.
  • Due to its low affinity for the target, an increase in avidity is required to ensure sufficient binding to the cell surface
  • Combining Fab-Streps with a Strep-Tactin™ backbone leads to the formation of complexes that stably bind to cells.
  • Fab-Streps were developed for Traceless Affinity Cell Selection (Fab-TACS™), which is based on our Strep-tag™ technology.
Specifications
Specifications
Lyophilized
Fluorescent Cell Staining & Sorting, Magnetic or Affinity Chromatographic Cell Isolation
Mouse
Murine CD8
Special Offers
For Research Use Only.