Learn More
Motic™ CCIS™ Plan Achromatic Strain-free Objectives
Plan Achromatic strain-free objectives for use with TEC POL Microscopes.
Brand: Motic™ 1101001707231
Additional Details : Weight : 0.12000kg
Description
- For use with Motic™ Panthera TEC POL Microscopes
- UC PL P Plan Achromatic strain-free objectives
Specifications
Microscope Objective | |
Plan Achromatic | |
0.8 |
Motic™ TEC POL Microscopes | |
60X | |
0.35 mm |