Learn More
Bochem™ Cast Iron H-frame Complete
Product Code. 10152551
Change view
Click to view available options
Length (Metric):
500 mm
Height (Metric):
800 mm
Weight (Metric):
6.5 kg
Includes:
Aluminum rods 12mm diameter and boss heads
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Height (Metric) Weight (Metric) Includes Unit Size
10152551 500 mm 800 mm 6.5 kg Aluminum rods 12mm diameter and boss heads Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10152551 Supplier Bochem™ Supplier No. 5641
  • ONLINE EXCLUSIVE PRICE
    350.00€
    290.14€ / Pack of 1
    Save 59.86€
    17% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Includes complete frame. BOCHEM™ Cast Iron H-frame Complete includes aluminum rods and bossheads.
  • Includes aluminum rods and bossheads
  • Cast iron is green varnished
Specifications
Material Cast Iron
Height (Metric) 800 mm
Length (Metric) 500 mm
Includes Aluminum rods 12mm diameter and boss heads
Product Type H-frame Complete
Quantity 1 Ea.
Weight (Metric) 6.5 kg