Learn More
Thermo Scientific™ Barnstead™ Water System Cartridges
Product Code. 11337121 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
High purity water cartridge
Ion exchange cartridge
Ultrapure polisher cartridge
For Use With (Equipment):
GenPure GenPure Pro, GenPure xCAD, LabTower EDI
MicroPure
Pacific TII
Smart2Pure
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description For Use With (Equipment) Unit Size
11337121 Ultrapure polisher cartridge MicroPure Each
11397151 High purity water cartridge Pacific TII Each
11357121 Ion exchange cartridge Smart2Pure Each
11367121 Ultrapure polisher cartridge Smart2Pure Each
15390471 Ultrapure polisher cartridge GenPure GenPure Pro, GenPure xCAD, LabTower EDI Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11337121 Supplier Thermo Scientific™ Supplier No. 09.1006
  • ONLINE EXCLUSIVE PRICE
    685.00€
    616.00€ / Pack of 1
    Save 69.00€
    10% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Maintain your Thermo Scientific™ Barnstead™ point of use water purification system with these replacement cartridges.
Maintain your Thermo Scientific™ Barnstead™ point of use water purification systems with replacement Thermo Scientific™ Barnstead™ Cartridges.
Choose the appropriate part number for your system. These cartridges are for use with the following Barnstead point of use water purification systems:
  • MicroPure™
  • GenPure™
  • GenPure™ Pro
  • GenPure™ xCAD
  • Smart2Pure™
  • LabTower™ EDI
  • Pacific™ TII
TRUSTED_SUSTAINABILITY
Specifications
Description Ultrapure polisher cartridge
For Use With (Equipment) MicroPure
Type Cartridge, Ultrapure Polisher