Learn More
Carl Zeiss™ Replacement Bulbs for Microscopes and Fiber Optic Power Supplies
For Carl Zeiss Microscopes and Fiber Optic Power Supplies
Brand:  Carl Zeiss™ 381619-0000-000
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 10296982
  • 651.90€ / Each
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Description
Description
  • Accessory for use with Carl Zeiss Stereomicrscopes
  • Halogen bulb
Specifications
Specifications
Mercury Burner HBO
Axiostar Plus Student, Axiostar Plus ICS
110 V
50 w