Learn More
Heathrow Scientific Cardboard Cryogenic Vial Box Partition 64 Well, Array 8 x 8, White
The Cardboard Partition is for organization of small samples and tubes.
Brand: Heathrow Scientific HS2860F
Additional Details : Weight : 0.30000kg
Description
- Partition has an 8 x 8 array
- 1.5/2.0 mL Tubes
- 64 Wells (15 mm dia.)
Specifications
64-Wells | |
White | |
Tubes | |
5.10 in. | |
5.10 in. | |
1.50 in. | |
15 mm |
Cardboard Cryogenic Vial Box Partition | |
Cardboard | |
8 x 8 | |
12.95 cm | |
12.95 cm | |
3.81 cm |