Learn More
Azpack™ Carbon Steel Razor Blades
Product Code. 11904325
Change view
Click to view available options
Size:
38.1 x 19 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11904325 38.1 x 19 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11904325 Supplier Azpack™ Supplier No. 11904325
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
In Stock
Add to basket
Edge
Add to basket
Packaged in drawer box merchandiser. Azpack™ Carbon Steel Razor Blades are individually wrapped in paper cardstock for safety.
  • Carbon steel
  • Regular duty
  • Single-edged blade
  • Contains aluminum spine
  • Individually wrapped
Specifications
Blade Material Carbon Steel
Product Type Razor Blade
Size 38.1 x 19 mm
Thickness (Metric) 0.22 mm
Quantity 10/Pk.