Learn More
Thermo Scientific™ ABgene™ Cap strip, for PCR tubes, strips of eight flat caps
Brand:  Thermo Scientific™ ABgene™ AB-0784-RMP340
Additional Details : Weight : 0.01000kg
Product Code. 15353139
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • Compatible with 96-well PCR plates and standard and low profile 0.2mL strip tubes
  • Certified DNase, RNase and human DNA free
  • Domed and flat available
  • Ultra clear model available for QPCR and fluorescence applications
  • Compatible with 96-well PCR plates and standard and low profile strip tubes
Cap style: Flat