Learn More
Motic™ C-Mount Camera Adapter for SMZ168 Stereo Zoom Trinocular Microscope Chip Sensor
Mount a digital C-mount camera with 1/3 in., 1/2 in., or 2/3 in. on board chip on your SMZ168 Trinocular
Brand: Motic™ SW0113N6
Additional Details : Weight : 0.80000kg
Description
- For use with SMZ168 Stereo Zoom Trinocular Microscope:
- SW0113N6: Mount a digital C-mount camera with 1/2 in. and 2/3 in. on board chip for 0.5X magnification power
- SW0113N7: Mount a digital C-mount camera with 1/3 in. on board chip for 0.45X magnification power
Specifications
Camera Adapter | |
12.7, 16.9 mm | |
0.5X |
1/2, 2/3 in. | |
SMZ168 Stereo Zoom Microscope | |
1 Ea. |