Learn More
YSI™ Cables and DO/Temp/Conductivity Probes for 556 Multi-Probe System
Handheld multiprobe meter
Brand: YSI™ 556310
Additional Details : Weight : 0.80000kg
Description
DO/Temperature/Conductivity Probe and Cable. Required to measure dissolved oxygen, temperature, conductivity.
Specifications
32 ft. | |
Temperature/conductivity sensor | |
YSI model 556 handheld multi probe meter | |
Measures DO, temperature, conductivity |
10 m | |
1 Year | |
DO/Temperature/Conductivity |