Learn More
IBA Lifesciences Buffer IS for Streptamers
IBA Lifesciences Buffer IS for preparation of wash buffer for the use in Streptamer technology.
Brand:  IBA Lifesciences 6-5602-050
Additional Details : Weight : 0.08500kg
Product Code. 16451485
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • 10X Concentrated
  • Contains 5 % BSA (w/v) in 10X Phosphate Buffered Saline (PBS)
Specifications
Specifications
50 mL
7.3 to 7.5
Washing Buffer for Streptamer™ Technology
10X
2°C to 8°C
Wash Buffer