Learn More
Witeg™ Buechner Filter Funnels
For use in general filtration applications in the laboratory.
Brand:  Witeg™ 2 367 053
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11857810
  • 31.40€ / Each
Estimated Shipment: 24-05-2024
Description
  • Made from borosilicate glass
  • Multiple capacities available
  • Multiple disc sizes and stem lengths
Specifications
Specifications
Buechner filter funnel
50 mL
Filtration
P3
35 mm
Borosilicate Glass
10 mm