Learn More
Thermo Scientific™ Buckets or Lids for Thermo Scientific™ TX-150 Swinging Bucket Rotor
Product Code. 15152693 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Bucket
Bucket Lid
Capacity:
4 x 145 mL
8 x 50 mL
Not Applicable
Unit Size:
Each
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Capacity Unit Size
15152693 Bucket Lid Not Applicable Each
15165233 Bucket 4 x 145 mL Pack of 4
15145253 Bucket 8 x 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15152693 Supplier Thermo Scientific™ Supplier No. 75005707
  • ONLINE EXCLUSIVE PRICE
    289.00€
    276.00€ / Pack of 1
    Save 13.00€
    4% Off
In Stock
Add to basket
Add to basket
Round and conical buckets offered in sets of four
Round and conical buckets or lids offered in sets of four
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Round Buckets
Capacity Not Applicable
Product Type Bucket Lid
Quantity Set of 4
Description ClickSeal Biocontainment Lids for Round Buckets