Learn More
Paragon Scientific™ BS/U/M Miniature Viscometer for Transparent Liquids
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
Brand:  Paragon Scientific™ MINIU05NC
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 16010250
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Specifications
20 to 100 cSt
0.1
ASTM D 445, D 446 and ISO 3104, 3105
No
D 445, D 446
M5
Special Offers
4 week lead-time