Learn More
Fisherbrand™ Ion Selective Electrode
Product Code. 11715648 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Membrane:
Combination
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Membrane Unit Size
11715648 Combination Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11715648
  • ONLINE EXCLUSIVE PRICE
    477.00€
    302.17€ / Pack of 1
    Save 174.83€
    36% Off
View all promotions
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
Combined ion-sensitive bromide electrode with solid membrane interchangeable flat surface is ideally delivered with membrane, electrolytic solution and gasket.
  • Combined ion-sensitive bromide electrode with solid membrane interchangeable flat surface
  • The ion to be detected passes through the membrane generating a potential difference proportional to the ionic concentration
  • Analysis time typically less than one minute
  • Sample preparation steps such as filtering or distillation rarely necessary
Specifications
Description Bromide Ion Selective Electrode
pH Range 2 to 14
Membrane Combination