missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates 96-Well, immunoGrade-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:Without Lid
Brand: BRAND™ 781732
85.00 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 1.50000kg
Description
- High-binding
- Optimized for the immobilization of IgG
- Offers the highest binding capacity for molecules with mixed hydrophilic and hydrophobic regions
- The surface of choice for the majority of standard ELISAs
- Suitable for solid phase immunoassays
- Comparable to high-binding plates from other manufacturers
- Free from endotoxins, DNase, DNA, RNase, non-cytotoxic
- Non-sterile
Specifications
Without Lid | |
Polystyrene | |
Non-sterile | |
330 μL | |
Yes | |
Clear |
Black, Clear Bottom | |
96 | |
immunoGrade | |
Flat Bottom (F-well) | |
Yes |