Learn More
Brady™ BradyJet™ J2000 Polyester Tags
Brady™ BradyJet™ J2000 Polyester Tags are ideal for identification applications.
Brand: Brady™ 150030
Additional Details : Weight : 0.45000kg
Description
- Material is a thick polyester tagstock material, with an inkjet receptive vinyl adhered
- Ideal for use in general facility labeling, including accident prevention and confined space tags
- Compliant with OSHA standard 1910.145 for accident prevention tags
- Compliant with OSHA standard 1910.147 for lockout tagout
Specifications
Acrylic | |
Matte | |
Polyester | |
BradyJet™ | |
Rectangle | |
-40°C to +60°C |
White | |
Roll | |
Arc Flash Labels, GHS and Chemical Labels, Lockout Tagout Labeling, Pipe and Valve Marking, Safety and Compliance ID, Visual Workplace Labeling, Warehouse Marking | |
100/Pk. | |
−40 to 140°F | |
5.75 x 3.25 in. |