Learn More
Brady™ BBP™ Repositional Vinyl Film
BBP™ Repositional Vinyl Film (B-581) is a durable, flexible, high gloss material ideal for indoor marking where frequent repositioning is required.
115.26€ - 415.23€
Specifications
| Adhesive Type | Acrylic, Repositionable, Pressure-sensitive |
|---|---|
| Finish | Gloss |
| Format | Film |
| Material | Vinyl |
| Sterility | Non-sterile |
| Product Code | Brand | Color | Width (English) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Color | Width (English) | Price | Quantity & Availability | ||||
|
11706465
|
Brady™
118003 |
White | 4 in. |
492.00€ 415.23€ / Pack of 1 Save 76.77€ 15% Off |
In Stock
Log in to see stock. |
||||
|
11716465
|
Brady™
118004 |
White | 2.25 in. |
328.00€ 276.78€ / Pack of 1 Save 51.22€ 15% Off |
In Stock
Log in to see stock. |
||||
|
11736465
|
Brady™
118006 |
White | 0.5 in. |
138.00€ 115.26€ / Pack of 1 Save 22.74€ 16% Off |
Estimated Shipment: 09-12-2026
Log in to see stock. |
||||
|
11756465
|
Brady™
118008 |
Yellow | 2.25 in. |
328.00€ 276.78€ / Pack of 1 Save 51.22€ 15% Off |
In Stock
Log in to see stock. |
||||
|
11726465
|
Brady™
118005 |
White | 1.12 in. |
220.00€ 184.51€ / Pack of 1 Save 35.49€ 16% Off |
In Stock
Log in to see stock. |
||||
|
11766465
|
Brady™
118009 |
Yellow | 1.12 in. |
220.00€ 184.51€ / Pack of 1 Save 35.49€ 16% Off |
In Stock
Log in to see stock. |
||||
|
11746465
|
Brady™
118007 |
Yellow | 4 in. |
492.00€ 415.23€ / Pack of 1 Save 76.77€ 15% Off |
In Stock
Log in to see stock. |
||||
|
11776465
|
Brady™
118010 |
Yellow | 0.5 in. |
138.00€ 115.26€ / Pack of 1 Save 22.74€ 16% Off |
Estimated Shipment: 09-12-2026
Log in to see stock. |
||||
Description
- Special aggressive adhesive stays in place but removes cleanly without leaving adhesive residue making it ideal for applications like 5S and inventory
- Vinyl film can be thermal transfer printed
- Average outdoor durability is 5 years
- Service temperature of -40°F to 180°F (-40°C to 82°C)
Specifications
| Acrylic, Repositionable, Pressure-sensitive | |
| Film | |
| Non-sterile | |
| BBP30, BBP31, BBP33, BBP35, and BBP37 printers | |
| 1 Roll | |
| -40°C to +82°C | |
| 100 ft. |
| Gloss | |
| Vinyl | |
| Ideal for temporary, Removable and repositionable labels, like 5S and lean inventory label | |
| 5 Years | |
| -40 to 180°F | |
| 0.005 in. | |
| 30.48 m |