Learn More
Velp™ Bosshead Double Clamp
Product Code. 10373230 Shop All Velp Products
Change view
Click to view available options
Type:
Boss Head Double Clamp
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10373230 Boss Head Double Clamp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10373230 Supplier Velp™ Supplier No. A00001301
In Stock
Add to basket
Edge
Add to basket
Velp™ Bosshead Double Clamp for use with Velp™ Overhead Stirrer and OV5 Homogenizers.
  • For use with Overhead Stirrer and OV5 Homogenizers
Specifications
Color Gray
For Use With (Equipment) VELP Overhead Stirrer and OV5 homogenizers
Quantity 1 Ea.
Type Boss Head Double Clamp
Material Cast Zinc, Aluminum