Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug
Product Code. 10731191
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Joint Number:
19/26 NS
29/32 NS
Pore Size:
2.5mm
4mm
6mm
Unit Size:
Pack of 2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Joint Number Pore Size unitSize
10731191 100 mL 19/26 NS 2.5mm Pack of 2
10102051 1000 mL 29/32 NS 6mm Pack of 2
10373371 100 mL 29/32 NS 2.5mm Pack of 2
12840453 250 mL 29/32 NS 4mm Pack of 2
10694192 500 mL 29/32 NS 4mm Pack of 2
10461711 50 mL 19/26 NS 2.5mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10731191 Supplier Lenz Laborglasinstrumente™ Supplier No. 04030137
Add to basket
Edge
Add to basket
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
  • With NS socket and PE stopper
  • Without scale, with PTFE plug
  • With screwthread retaining device
  • With inscription label
Specifications
Capacity (Metric) 100 mL
Material Borosilicate Glass
Joint Number 19/26 NS
Pore Size 2.5mm
Quantity 2Pack
Stopper Material Polyethylene