Learn More
Fisherbrand™ Glass Circle Coverslips
Product Code. p-8000283 Shop All Fisherbrand Products
Change view
Click to view available options
Thickness (Metric):
#1 (0.13 to 0.16 mm)
#1.5 (0.16 to 0.19 mm)
Diameter (Metric):
16 mm
19 mm
22 mm
Unit Size:
Pack of 100
Pack of 200
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Diameter (Metric) unitSize
12313138 #1 (0.13 to 0.16 mm) 16 mm Pack of 200
12323148 #1.5 (0.16 to 0.19 mm) 16 mm Pack of 100
12333138 #1 (0.13 to 0.16 mm) 22 mm Pack of 200
12323138 #1 (0.13 to 0.16 mm) 19 mm Pack of 200
11507323 #1.5 (0.16 to 0.19 mm) 22 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12313138
Only null left
Add to basket
Edge
Add to basket
Glass Circle Coverslips are available in two thicknesses and a variety of diameters. Packed in plastic boxes to prevent breakage.
These Glass Circle Coverslips are available in two thicknesses and a variety of diameters. Packed in plastic boxes to prevent breakage.
  • Manufactured from clear soda glass
  • Consistent in cut, thickness and dimensional accuracy
  • Free from blisters and flaws
Specifications
Product Type Coverslip
Shape Circle
Material Soda Lime Glass
Thickness (Metric) #1 (0.13 to 0.16 mm)
Diameter (Metric) 16 mm
Quantity 200/Pk.