Learn More
Robu™ Borosilicate Glass Micro-Filter Funnels
Brand: Robu™ 28043
Additional Details : Weight : 0.06460kg
Specifications
4 mL | |
Borosilicate Glass | |
Highly resistant to most chemicals such as solvents, acids and alkalis | |
3 |
6mm | |
55 mm | |
Unground joint | |
Micro Filter Funnel |