Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Filter Funnels
Lenz Laborglasinstrumente™ Filter funnels are ideal for separating solids from liquids via laboratory filtration.
54.96€ - 146.28€
Product Code | Brand | Capacity (Metric) | Porosity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Porosity | Price | Quantity & Availability | ||||
10767331
|
Lenz Laborglasinstrumente™
07195340 |
125 mL | 3 |
72.08€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10459942
|
Lenz Laborglasinstrumente™
07195158 |
500 mL | 1 |
134.62€
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
10652092
|
Lenz Laborglasinstrumente™
07195134 |
75 mL | 1 |
54.96€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10460133
|
Lenz Laborglasinstrumente™
07195458 |
500 mL | 4 |
146.28€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10577671
|
Lenz Laborglasinstrumente™
07195358 |
500 mL | 3 |
129.32€
Each |
In Stock
Log in to see stock. |
||||
Description
- Made from borosilicate glass
- Separates solids from liquids
Specifications
Borosilicate Glass | |
Filter funnel |