Learn More
SHOWA™ Blue Chemical Protection Gloves, Model CS700
Description
- A double-dipped nitrile coating provides chemical protection and ensures the gloves are highly durable to keep the wearer safe from harmful substances.
- The combination of a seamless liner and natural latex-free design is kind to the skin.
Specifications
Specifications
| Chlorinated | No |
| Color | Blue |
| Finger Thickness (English) | 45.6 mil |
| Finger Thickness (Metric) | 1.16 mm |
| Latex Free | Yes |
| Material | Nitrile, Polyester |
| Palm Thickness (English) | 45.6 mil |
| Palm Thickness (Metric) | 1.16 mm |
| Size | 8 |
| Thickness (English) | 45.6 mil |
| Show More |