Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Calibrated Density Bottles
Product Code. 10747981 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10747981 25 mL Each
10663561 10 mL Each
10693371 5 mL Each
10748171 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10747981 Supplier BRAND™ Supplier No. 43320
  • ONLINE EXCLUSIVE PRICE
    77.28€
    57.36€ / Pack of 1
    Save 19.92€
    25% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Calibrated density bottle made with Borosilicate glass that is ideal for use in most research, scientific, and other types of laboratory environments.
Gay-Lussac density bottles manufactured from borosilicate glass.
  • Conforms to DIN ISO 3507 standards
  • Calibrated to contain (TC, In).
  • Includes individual certificate.
  • Stopper NS 10/19 with capillary.
  • Top of stopper ground and polished.
  • The volume in cm³ is specified to a precision of 3 decimal places.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1Each
Type Gay-Lussac
Capacity (Metric) 25 mL
Material Borosilicate Glass
Product Type Pycnometer