Learn More
Fisherbrand™ Black Phenolic Caps
Black phenolic screw cap with poly-seal liner is manufactured from low density polyethylene. For use with various Fisherbrand molded glass bottles
Brand:  Fisherbrand™
Additional Details : Weight : 1.57500kg
 View more versions of this product
Product Code. 16164587
  • 290.00€ / Pack of 576
Estimated Shipment: 10-08-2022
Description
Unique cone liner design offers superior torque retention and excellent sealing characteristics
Specifications
Specifications
Black
Lined Cap
15-425
No
576
Black
Polyethylene Cone
Phenolic