Learn More
Bis(tetramethylcyclopentadienyl)chromium(II), 98+%, Thermo Scientific™
Catalog Number 11377199
Click to view available options
Quantity:
1 g
5 g
82066-37-3
C18H26Cr
294.402
MFCD01862451
XSSFNWHTVOECSF-UHFFFAOYSA-N
71431265
chromium(2+);1,2,3,5-tetramethylcyclopenta-1,3-diene
CC1[C-]=C(C(=C1C)C)C.CC1[C-]=C(C(=C1C)C)C.[Cr+2]
82066-37-3
C18H26Cr
294.402
MFCD01862451
XSSFNWHTVOECSF-UHFFFAOYSA-N
71431265
chromium(2+);1,2,3,5-tetramethylcyclopenta-1,3-diene
CC1[C-]=C(C(=C1C)C)C.CC1[C-]=C(C(=C1C)C)C.[Cr+2]
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Melting Point 106°C to 111°C
Quantity 1 g
Sensitivity Air and moisture sensitive
Formula Weight 294.41
Percent Purity ≥98%
Chemical Name or Material Bis(tetramethylcyclopentadienyl)chromium(II)