Learn More
Macherey-Nagel™ Bioanalysis MN 827 B Blotting Papers
Brand:  Macherey-Nagel™ 742128
Additional Details : Weight : 0.72000kg
 View more versions of this product
Product Code. 10003091
  • 53.10€ / Pack of 100
Estimated Shipment: 26-06-2024
Specifications
Blotting Paper
100 Pack
0.7 mm
160 x 160 mm