Learn More
Bethanechol chloride, 98%, Thermo Scientific™
A cholinergic receptor agonist
42.50€ - 131.00€
Chemical Identifiers
| CAS | 590-63-6 |
|---|---|
| Molecular Formula | C7H17ClN2O2 |
| Molecular Weight (g/mol) | 196.675 |
| MDL Number | MFCD00055224 |
| InChI Key | XXRMYXBSBOVVBH-UHFFFAOYSA-N |
| Synonym | bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone |
| PubChem CID | 11548 |
| ChEBI | CHEBI:3085 |
| IUPAC Name | 2-carbamoyloxypropyl(trimethyl)azanium;chloride |
| SMILES | CC(C[N+](C)(C)C)OC(=O)N.[Cl-] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15477129
|
Thermo Scientific Alfa Aesar
J65919.03 |
1g |
42.50€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15487129
|
Thermo Scientific Alfa Aesar
J65919.06 |
5g |
131.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C7H17ClN2O2 | |
| MFCD00055224 | |
| bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone | |
| CHEBI:3085 | |
| CC(C[N+](C)(C)C)OC(=O)N.[Cl-] |
Specifications
| 590-63-6 | |
| MFCD00055224 | |
| bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone | |
| CC(C[N+](C)(C)C)OC(=O)N.[Cl-] | |
| 196.675 | |
| CHEBI:3085 | |
| 98% |
| C7H17ClN2O2 | |
| 14,1185 | |
| XXRMYXBSBOVVBH-UHFFFAOYSA-N | |
| 2-carbamoyloxypropyl(trimethyl)azanium;chloride | |
| 11548 | |
| 196.68 | |
| Bethanechol chloride |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 209-686-8
missing translation for 'rtecsNumber' : BR5425000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.